Gene Transcripts

Gene Symbol
Gemin2
Entrez Gene ID
66603
Gene Name
gem (nuclear organelle) associated protein 2
Gene Aliases
1700012N19Rik, Sip1
Chromosome Location
Chr. 12: 59013393 - 59027988 on Build GRCm38
UniGene ID
Mm.35353
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025656.4 NP_079932.2 7 628
GenBank mRNA AK005928.1 - 7 630
AK007515.1 - 7 629
AK013414.1 - 7 585
BC053424.1 - 7 1057 6336522

View Full Product Details