Gene Transcripts

Gene Symbol
Clcnkb
Entrez Gene ID
56365
Gene Name
chloride channel Kb
Gene Aliases
CLCKB, Clc-Ka, Clck2, Clcnk1l
Chromosome Location
Chr. 4: 141404357 - 141416013 on Build GRCm38
UniGene ID
Mm.24882
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019701.2 NP_062675.2 12 1235
GenBank mRNA AF124847.1 - 12 1228
AK146876.1 - 11 1298
BC019983.1 - 12 1261 4188062

View Full Product Details