Gene Transcripts

Gene Symbol
Syn3
Entrez Gene ID
29130
Gene Name
synapsin III
Gene Aliases
Chromosome Location
Chr. 7: 23403896 - 23808602 on Build Rnor_6.0
UniGene ID
Rn.162920
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017109.1 NP_058805.1 4 646
GenBank mRNA AF056704.1 - 4 646

View Full Product Details