Gene Transcripts

Gene Symbol
Cd82
Entrez Gene ID
83628
Gene Name
Cd82 molecule
Gene Aliases
Kai1
Chromosome Location
Chr. 3: 82324344 - 82368846 on Build Rnor_6.0
UniGene ID
Rn.3022
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031797.2 NP_113985.2 8 815
XM_006234597.2 XP_006234659.1 8 889
XM_006234598.1 XP_006234660.1 8 703
XM_008762039.1 XP_008760261.1 7 608
GenBank mRNA AF049882.1 - 8 729
BC072498.1 - 8 815 7096826

View Full Product Details