Gene Transcripts

Gene Symbol
Cldn16
Entrez Gene ID
155268
Gene Name
claudin 16
Gene Aliases
Pcln1
Chromosome Location
Chr. 11: 77683942 - 77703232 on Build Rnor_6.0
UniGene ID
Rn.43852
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_131905.1 NP_571980.1 5 1443
GenBank mRNA AF333099.1 - 5 1443

View Full Product Details