Gene Transcripts

Gene Symbol
Slco4a1
Entrez Gene ID
171144
Gene Name
solute carrier organic anion transporter family, member 4a1
Gene Aliases
OATP-E, Slc21a12
Chromosome Location
Chr. 3: 175838994 - 175857985 on Build Rnor_6.0
UniGene ID
Rn.67081
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133608.2 NP_598292.2 2 534
GenBank mRNA AF239262.1 - 1 368
BC097285.1 - 2 534 7441748
BC097286.1 - 2 476 7444297

View Full Product Details