Gene Transcripts

Gene Symbol
Hspa4
Entrez Gene ID
266759
Gene Name
heat shock protein 4
Gene Aliases
Hsp110, Hsp70, irp94
Chromosome Location
Chr. 10: 38601624 - 38642441 on Build Rnor_6.0
UniGene ID
Rn.163092
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153629.1 NP_705893.1 2, 3 384
XM_006246232.1 XP_006246294.1 2, 3 428
GenBank mRNA AF077354.1 - 2, 3 384
FQ222433.1 - 3, 4 592

View Full Product Details