Gene Transcripts

Gene Symbol
Mybbp1a
Entrez Gene ID
60571
Gene Name
MYB binding protein (P160) 1a
Gene Aliases
DBP
Chromosome Location
Chr. 10: 59000401 - 59010789 on Build Rnor_6.0
UniGene ID
Rn.35696
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031668.1 NP_113856.1 5 430
XM_006246802.1 XP_006246864.1 6 674
GenBank mRNA U83590.1 - 5 430

View Full Product Details