Gene Transcripts

Gene Symbol
Kcnk15
Entrez Gene ID
156873
Gene Name
potassium channel, two pore domain subfamily K, member 15
Gene Aliases
Chromosome Location
Chr. 3: 160231914 - 160238132 on Build Rnor_6.0
UniGene ID
Rn.161865
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130813.1 NP_570826.1 2 1136
GenBank mRNA AF467250.1 - 2 1136

View Full Product Details