Gene Transcripts

Gene Symbol
Prokr2
Entrez Gene ID
192649
Gene Name
prokineticin receptor 2
Gene Aliases
Gpr73l1
Chromosome Location
Chr. 3: 125006210 - 125020847 on Build Rnor_6.0
UniGene ID
Rn.82760
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138978.1 NP_620434.1 2 703
XM_006235059.2 XP_006235121.1 4 1112
GenBank mRNA AY089975.1 - 2 703

View Full Product Details