Gene Transcripts

Gene Symbol
Gss
Entrez Gene ID
25458
Gene Name
glutathione synthetase
Gene Aliases
Chromosome Location
Chr. 3: 151076253 - 151106557 on Build Rnor_6.0
UniGene ID
Rn.1692
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012962.1 NP_037094.1 12 1183
XM_008762303.1 XP_008760525.1 12 1597
GenBank mRNA BC078700.1 - 11 1148
L38615.1 - 12 1183

View Full Product Details