Gene Transcripts

Gene Symbol
Snai2
Entrez Gene ID
25554
Gene Name
snail family zinc finger 2
Gene Aliases
Slug
Chromosome Location
Chr. 11: 90404421 - 90406730 on Build Rnor_6.0
UniGene ID
Rn.43117
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013035.1 NP_037167.1 2 470
GenBank mRNA AF497973.1 - 2 470
U97061.1 - 1 404

View Full Product Details