Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286350.1 | NP_001273279.1 | 3 | 975 | |
NM_214462.4 | NP_999627.2 | 4 | 1498 | ||
NR_104425.1 | 2 | 507 | |||
XM_011535507.2 | XP_011533809.1 | 4 | 1260 | ||
GenBank mRNA | AF318336.1 | - | 4 | 1466 | |
BC111764.1 | - | 2 | 463 | ||
BC111790.1 | - | 3 | 931 |