Gene Transcripts

Gene Symbol
Tspan3
Entrez Gene ID
300733
Gene Name
tetraspanin 3
Gene Aliases
Tm4sf8
Chromosome Location
Chr. 8: 60806848 - 60830448 on Build Rnor_6.0
UniGene ID
Rn.998
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005547.1 NP_001005547.1 4 447
GenBank mRNA BC082102.1 - 4 447

View Full Product Details