Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199691.1 | NP_001186620.1 | 8 | 1242 | |
GenBank mRNA | AK309126.1 | - | 3 | 685 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001128142.1 | NP_001121614.1 | 2 | 657 | |
NR_103483.1 | 2 | 657 | |||
GenBank mRNA | AK292665.1 | - | 2 | 609 | |
AK292699.1 | - | 2 | 657 | ||
AK296476.1 | - | 3 | 743 | ||
AL050062.1 | - | 2 | 601 | ||
BC010936.2 | - | 2 | 593 | ||
BX537497.1 | - | 1 | 1351 | ||
CR533493.1 | - | 2 | 581 |