Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017414.3 | NP_059110.2 | 3 | 559 | |
XM_006724074.2 | XP_006724137.1 | 3 | 630 | ||
GenBank mRNA | AF176642.2 | - | 2 | 241 | |
AJ243526.1 | - | 3 | 424 | ||
AK313385.1 | - | 3 | 380 | ||
AL136690.1 | - | 3 | 380 | ||
BC014896.1 | - | 3 | 365 | 3910720 | |
BI752849.1 | - | 3 | 455 | ||
BT006835.1 | - | 2 | 221 | ||
CR457216.1 | - | 2 | 221 | ||
CT841507.1 | - | 2 | 287 |