Gene Transcripts

Gene Symbol
Plscr4
Entrez Gene ID
300900
Gene Name
phospholipid scramblase 4
Gene Aliases
Pls4
Chromosome Location
Chr. 8: 99880073 - 99917733 on Build Rnor_6.0
UniGene ID
Rn.37802
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012000.1 NP_001012000.1 7 1035
GenBank mRNA AY548832.1 - 7 1035

View Full Product Details