Gene Transcripts

Gene Symbol
Mllt11
Entrez Gene ID
295264
Gene Name
myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 11
Gene Aliases
RGD1305525
Chromosome Location
Chr. 2: 196392936 - 196402106 on Build Rnor_6.0
UniGene ID
Rn.144837
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013912.1 NP_001013934.1 2 549
GenBank mRNA BC087583.1 - 2 549
BC094215.1 - 2 549 7318415

View Full Product Details