Gene Transcripts

Gene Symbol
Spag8
Entrez Gene ID
433700
Gene Name
sperm associated antigen 8
Gene Aliases
MH-SPAG8
Chromosome Location
Chr. 4: 43651330 - 43654124 on Build GRCm38
UniGene ID
Mm.331569
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007463.1 NP_001007464.1 2 123
NM_001290462.1 NP_001277391.1 2 123
GenBank mRNA AK132968.1 - 2 124
AY769082.1 - 2 123
BC150868.1 - 2 165 9087779

View Full Product Details