Gene Transcripts

Gene Symbol
KRBOX4
Entrez Gene ID
55634
Gene Name
KRAB box domain containing 4
Gene Aliases
ZNF673
Chromosome Location
Chr. X: 46447189 - 46474639 on Build GRCh38
UniGene ID
Hs.632800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129898.1 NP_001123370.1 6 2023
NM_001129899.1 NP_001123371.1 7 2067
NM_001129900.1 NP_001123372.1 7 2052
NM_017776.2 NP_060246.2 6 2008
GenBank mRNA AK000351.1 - 6 1875
AK096413.1 - 6 1917
AK097159.1 - 1 2893

View Full Product Details