Gene Transcripts
- Gene Symbol
- Sdccag3
- Entrez Gene ID
- 306322
- Gene Name
- serologically defined colon cancer antigen 3
- Gene Aliases
- Chromosome Location
- Chr. 3: 3827498 - 3834219 on Build Rnor_6.0
- UniGene ID
- Rn.110254
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100360061
- Entrez Gene ID
- 100360061
- Gene Name
- mCG18732-like
- Gene Aliases
- Chromosome Location
- Chr. 6: 82982303 - 82984185 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100363321
- Entrez Gene ID
- 100363321
- Gene Name
- serologically defined colon cancer antigen 3
- Gene Aliases
- Chromosome Location
- Chr. 16: 18379595 - 18381631 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100909786
- Entrez Gene ID
- 100909786
- Gene Name
- serologically defined colon cancer antigen 3 homolog
- Gene Aliases
- Chromosome Location
- Chr. 6: 82908401 - 82910283 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- RGD1562289
- Entrez Gene ID
- 501516
- Gene Name
- similar to RIKEN cDNA C330016H24
- Gene Aliases
- Chromosome Location
- Chr. X: 7049662 - 7051625 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_576917.5 |
XP_576917.1
|
1 |
1131 |
|
View Full Product Details