Gene Transcripts

Gene Symbol
MUC12
Entrez Gene ID
10071
Gene Name
mucin 12, cell surface associated
Gene Aliases
MUC-11, MUC-12, MUC11
Chromosome Location
Chr. 7: 100969623 - 101018949 on Build GRCh38
UniGene ID
Hs.489355
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164462.1 NP_001157934.1 2 2892
GenBank mRNA AF147791.1 - 1 1395

View Full Product Details