Gene Transcripts
- Gene Symbol
- CASP1P2
- Entrez Gene ID
- 440067
- Gene Name
- caspase 1 pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 11: 105062602 - 105071530 on Build GRCh38
- UniGene ID
- Hs.446758
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_131905.1 |
|
2 |
112 |
|
GenBank mRNA |
CB985891.1 |
- |
2 |
115 |
|
View Full Product Details