Gene Transcripts

Gene Symbol
LOC283278
Entrez Gene ID
283278
Gene Name
pleckstrin homology domain-containing family A member 6
Gene Aliases
Chromosome Location
Chr. 11: 16778935 - 16787192 on Build GRCh38
UniGene ID
Hs.729280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017018657.1 XP_016874146.1 1 109
XM_017018658.1 XP_016874147.1 2 97

View Full Product Details