Gene Transcripts
- Gene Symbol
- TEX41
- Entrez Gene ID
- 401014
- Gene Name
- testis expressed 41 (non-protein coding)
- Gene Aliases
- DKFZp686O1327, LINC00953
- Chromosome Location
- Chr. 2: 144667967 - 145076729 on Build GRCh38
- UniGene ID
- Hs.742290
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033870.2 |
|
5 |
677 |
|
GenBank mRNA |
BC043549.1 |
- |
5 |
618 |
|
BX648102.1 |
- |
4 |
522 |
|
View Full Product Details