Gene Transcripts

Gene Symbol
Olr753
Entrez Gene ID
405237
Gene Name
olfactory receptor 753
Gene Aliases
Chromosome Location
Chr. 3: 102553317 - 102554234 on Build Rnor_6.0
UniGene ID
Rn.142770
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000921.1 NP_001000921.1 1 713

View Full Product Details