Gene Transcripts

Gene Symbol
Olfr531
Entrez Gene ID
258955
Gene Name
olfactory receptor 531
Gene Aliases
MOR251-3
Chromosome Location
Chr. 7: 140400130 - 140401044 on Build GRCm38
UniGene ID
Mm.223350
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146953.1 NP_667164.1 1 505
GenBank mRNA BC104052.1 - 1 505
BC104053.1 - 1 505 40040242

View Full Product Details