Gene Transcripts

Gene Symbol
Olfr504
Entrez Gene ID
258163
Gene Name
olfactory receptor 504
Gene Aliases
MOR40-15, MOR40-7P
Chromosome Location
Chr. 7: 108564837 - 108565793 on Build GRCm38
UniGene ID
Mm.377367
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011858.1 NP_001011858.1 1 352
GenBank mRNA BC104310.1 - 1 352

View Full Product Details