Gene Transcripts

Gene Symbol
Crim1
Entrez Gene ID
50766
Gene Name
cysteine rich transmembrane BMP regulator 1 (chordin like)
Gene Aliases
AU015004
Chromosome Location
Chr. 17: 78200185 - 78376592 on Build GRCm38
UniGene ID
Mm.311912
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015800.3 NP_056615.1 3 1258
GenBank mRNA AF168680.1 - 3 710
AK053197.1 - 1 201
BC100348.1 - 3 1043 30521148

View Full Product Details