Gene Transcripts
- Gene Symbol
- OTOA
- Entrez Gene ID
- 146183
- Gene Name
- otoancorin
- Gene Aliases
- CT108, DFNB22
- Chromosome Location
- Chr. 16: 21663857 - 21771468 on Build GRCh38
- UniGene ID
- Hs.408336
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161683.1 |
NP_001155155.1
|
8, 9 |
1185 |
|
NM_144672.3 |
NP_653273.3
|
12, 13 |
1329 |
|
NM_170664.2 |
NP_733764.1
|
3, 4 |
423 |
|
XM_011545747.2 |
XP_011544049.1
|
13, 14 |
1708 |
|
XM_011545748.2 |
XP_011544050.2
|
2, 3 |
702 |
|
XM_017022950.1 |
XP_016878439.1
|
13, 14 |
1432 |
|
GenBank mRNA |
AK093062.1 |
- |
3, 4 |
423 |
|
AK125840.1 |
- |
8, 9 |
1185 |
|
AK302733.1 |
- |
2, 3 |
485 |
|
BC129992.1 |
- |
12, 13 |
1326 |
|
BC129993.1 |
- |
12, 13 |
1333 |
40128985 |
View Full Product Details