Gene Transcripts

Gene Symbol
THAP8
Entrez Gene ID
199745
Gene Name
THAP domain containing 8
Gene Aliases
Chromosome Location
Chr. 19: 36034984 - 36054785 on Build GRCh38
UniGene ID
Hs.350209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152658.2 NP_689871.1 4 1379
XM_005258623.2 XP_005258680.1 4 1188
XM_011526604.2 XP_011524906.1 4 1388
GenBank mRNA AK057453.1 - 5 1235
AK093048.1 - 4 1380
AK296640.1 - 4 873
AK297630.1 - 3 689
BC046207.1 - 4 928 6176484
BC072416.1 - 4 1019 6169558
JQ410992.1 - 4 1256

View Full Product Details