Gene Transcripts

Gene Symbol
Ak4
Entrez Gene ID
29223
Gene Name
adenylate kinase 4
Gene Aliases
AK 4, Ak3l1, Ak3l2
Chromosome Location
Chr. 5: 120250306 - 120309153 on Build Rnor_6.0
UniGene ID
Rn.1086
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017135.3 NP_058831.1 1 80
XM_006238445.2 XP_006238507.1 2 385
GenBank mRNA BC087024.1 - 1 132
D87809.1 - 1 80

View Full Product Details