Gene Transcripts
- Gene Symbol
- Gm9776
- Entrez Gene ID
- 328309
- Gene Name
- predicted gene 9776
- Gene Aliases
- A530089L17, ENSMUSG00000042857
- Chromosome Location
- Chr. 13: 94356747 - 94358923 on Build GRCm38
- UniGene ID
- Mm.174191
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK041197.1 |
- |
1 |
944 |
|
AK141947.1 |
- |
|
863 |
|
AK162882.1 |
- |
1 |
970 |
|
View Full Product Details