Gene Transcripts

Gene Symbol
Gm5486
Entrez Gene ID
433024
Gene Name
predicted gene 5486
Gene Aliases
EG433024
Chromosome Location
Chr. 16: 49929454 - 49972476 on Build GRCm38
UniGene ID
Mm.328170
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK139943.1 - 954

View Full Product Details