Gene Transcripts

Gene Symbol
Rnf166
Entrez Gene ID
68718
Gene Name
ring finger protein 166
Gene Aliases
1110031E24Rik, AW555115, Zfp313l
Chromosome Location
Chr. 8: 122466145 - 122476064 on Build GRCm38
UniGene ID
Mm.34356
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033142.2 NP_001028314.1 6 1695
GenBank mRNA AK004000.1 - 1 322
AK079883.1 - 6 1682
AK151813.1 - 6 1695
AK209147.1 - 1 85

View Full Product Details