Gene Transcripts

Gene Symbol
Fam151b
Entrez Gene ID
73942
Gene Name
family with sequence similarity 151, member B
Gene Aliases
4930405M20Rik
Chromosome Location
Chr. 13: 92449620 - 92484290 on Build GRCm38
UniGene ID
Mm.85935
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163627.1 NP_001157099.1 2 119
GenBank mRNA AK015100.1 - 2 102
AK162422.1 - 2 103
BY202398.1 - 2 120

View Full Product Details