Gene Transcripts

Gene Symbol
Adam17
Entrez Gene ID
57027
Gene Name
ADAM metallopeptidase domain 17
Gene Aliases
TACE
Chromosome Location
Chr. 6: 43400525 - 43448280 on Build Rnor_6.0
UniGene ID
Rn.144585
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020306.2 NP_064702.1 3 556
GenBank mRNA AJ012603.1 - 3 558

View Full Product Details