Gene Transcripts

Gene Symbol
Rhag
Entrez Gene ID
65207
Gene Name
Rh-associated glycoprotein
Gene Aliases
Rh50
Chromosome Location
Chr. 9: 23465190 - 23493081 on Build Rnor_6.0
UniGene ID
Rn.44480
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023022.2 NP_075411.2 1 230
GenBank mRNA AB015194.1 - 1 203
AF531097.1 - 1 230

View Full Product Details