Gene Transcripts
- Gene Symbol
- Il12a
- Entrez Gene ID
- 84405
- Gene Name
- interleukin 12A
- Gene Aliases
- Chromosome Location
- Chr. 2: 165076945 - 165083996 on Build Rnor_6.0
- UniGene ID
- Rn.207199
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053390.1 |
NP_445842.1
|
2, 3 |
188 |
|
GenBank mRNA |
AF177031.1 |
- |
2, 3 |
188 |
|
View Full Product Details