Gene Transcripts

Gene Symbol
Il12a
Entrez Gene ID
84405
Gene Name
interleukin 12A
Gene Aliases
Chromosome Location
Chr. 2: 165076945 - 165083996 on Build Rnor_6.0
UniGene ID
Rn.207199
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053390.1 NP_445842.1 6 417
GenBank mRNA AF177031.1 - 6 417

View Full Product Details