Gene Transcripts

Gene Symbol
Fut9
Entrez Gene ID
84597
Gene Name
fucosyltransferase 9 (alpha (1,3) fucosyltransferase)
Gene Aliases
Chromosome Location
Chr. 5: 40032794 - 40237591 on Build Rnor_6.0
UniGene ID
Rn.212206
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053465.1 NP_445917.1 3 958
XM_008763598.1 XP_008761820.1 2 1013
GenBank mRNA AB049819.1 - 3 958
AF345993.1 - 2 661

View Full Product Details