Gene Transcripts
- Gene Symbol
- Rab3b
- Entrez Gene ID
- 81755
- Gene Name
- RAB3B, member RAS oncogene family
- Gene Aliases
- Chromosome Location
- Chr. 5: 128501787 - 128568170 on Build Rnor_6.0
- UniGene ID
- Rn.3788
- Species
- Rattus norvegicus
- Gene Symbol
- Rab3d
- Entrez Gene ID
- 140665
- Gene Name
- RAB3D, member RAS oncogene family
- Gene Aliases
- Chromosome Location
- Chr. 8: 22920238 - 22929361 on Build Rnor_6.0
- UniGene ID
- Rn.9822
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
S68807.1 |
- |
|
121 |
|
View Full Product Details