Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021600.3 | NP_067611.2 | 10 | 1275 | |
GenBank mRNA | AK081344.1 | - | 9 | 1261 | |
BC052153.1 | - | 10 | 1220 | 6336578 | |
BC141376.1 | - | 10 | 1203 | 9055989 | |
BC141387.1 | - | 10 | 1203 | 9056000 | |
K02582.1 | - | 10 | 1186 | ||
L10076.1 | - | 10 | 1235 |