Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026464.2 | NP_080740.2 | 2 | 270 | |
GenBank mRNA | AK018469.1 | - | 2 | 251 | |
AK075958.1 | - | 2 | 270 | ||
AK146093.1 | - | 2 | 289 | ||
BC064750.1 | - | 2 | 230 | ||
BC131932.1 | - | 2 | 264 | 40130209 | |
BC131934.1 | - | 2 | 264 | 40130211 |