Gene Transcripts

Gene Symbol
Vmn1r41
Entrez Gene ID
113857
Gene Name
vomeronasal 1 receptor 41
Gene Aliases
V1ra12, V1rb9
Chromosome Location
Chr. 6: 89740888 - 89747414 on Build GRCm38
UniGene ID
Mm.377154
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053230.2 NP_444460.2 2 745

View Full Product Details