Gene Transcripts

Gene Symbol
Card14
Entrez Gene ID
170720
Gene Name
caspase recruitment domain family, member 14
Gene Aliases
Bimp2, CARMA2
Chromosome Location
Chr. 11: 119307096 - 119345375 on Build GRCm38
UniGene ID
Mm.130832
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130886.3 NP_570956.1 3 418
GenBank mRNA AF363457.1 - 3 397
AK086176.1 - 4 667
BC029102.1 - 3 418 3979883

View Full Product Details