Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_139309.4 | NP_647470.1 | 1, 2 | 224 | |
GenBank mRNA | AB077383.1 | - | 2, 3 | 408 | |
AJ511807.1 | - | 2, 3 | 244 | ||
AK085931.1 | - | 1, 2 | 225 | ||
AK144743.1 | - | 2, 3 | 253 |