Gene Transcripts

Gene Symbol
Otoa
Entrez Gene ID
246190
Gene Name
otoancorin
Gene Aliases
Chromosome Location
Chr. 7: 121083438 - 121163470 on Build GRCm38
UniGene ID
Mm.221073
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139310.1 NP_647471.1 2, 3 171
GenBank mRNA AK157904.1 - 2, 3 203
AK158092.1 - 2, 3 288
AY055122.1 - 2, 3 171

View Full Product Details