Gene Transcripts

Gene Symbol
Fktn
Entrez Gene ID
246179
Gene Name
fukutin
Gene Aliases
D830030O17Rik, Fcmd
Chromosome Location
Chr. 4: 53714107 - 53765785 on Build GRCm38
UniGene ID
Mm.247210
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139309.4 NP_647470.1 9 1956
GenBank mRNA AB077383.1 - 10 2140
AK052357.1 - 12 2385
AK085931.1 - 9 1957
AK144743.1 - 9 1758
BC017538.1 - 10 1972 4506804

View Full Product Details