Gene Transcripts

Gene Symbol
Heatr1
Entrez Gene ID
217995
Gene Name
HEAT repeat containing 1
Gene Aliases
AA517551, B130016L12Rik, BC019693
Chromosome Location
Chr. 13: 12395375 - 12438893 on Build GRCm38
UniGene ID
Mm.171186
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144835.4 NP_659084.3 39, 40 5782
GenBank mRNA AK032134.1 - 4, 5 707
AK132192.1 - 1456
BC019693.1 - 3, 4 421

View Full Product Details